Anti-PCP4 Rabbit Polyclonal Antibody
New Product
ELISA:1:2000-1:10000, WB:1:500-1:2000, IHC:1:5-1:20
Type:
Antigen: PCP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77711-230
- 77711-229
- Antigen symbol:
- PCP4
- PCP4
- Antigen synonyms:
- Brain specific polypeptide PEP19|Purkinje cell protein 4|Pcp4|PCP4_HUMAN|Brain-specific antigen PCP-4|Brain specific antigen PCP 4|OTTHUMP00000109191|Brain specific polypeptide PEP 19|PEP19|Brain-specific polypeptide PEP-19
- Brain specific polypeptide PEP19|Purkinje cell protein 4|Pcp4|PCP4_HUMAN|Brain-specific antigen PCP-4|Brain specific antigen PCP 4|OTTHUMP00000109191|Brain specific polypeptide PEP 19|PEP19|Brain-specific polypeptide PEP-19
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P48539
- P48539
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human PCP4
- Fusion protein of Human PCP4
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together