Anti-NDUFS8 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:25-1:100
Type:
Antigen: NDUFS8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77712-719
- 77712-720
- Antigen symbol:
- NDUFS8
- NDUFS8
- Antigen synonyms:
- TYKY subunit|Complex I-23kD|NADH dehydrogenase [ubiquinone] iron-sulfur protein 8|Complex I 23kD|CI-23kD|NDUFS8|NADH-ubiquinone oxidoreductase 23 kDa subunit|EC 1.6.5.3|mitochondrial|NADH ubiquinone oxidoreductase 23 kDa subunit|EC 1.6.99.3|mitochondrial precursor|Human mitochondrial NADH dehydrogenase ubiquinone Fe S protein 8|CI 23kD|Ubiquinoneiron sulfur protein 8|NDUS8_HUMAN|TYKY
- TYKY subunit|Complex I-23kD|NADH dehydrogenase [ubiquinone] iron-sulfur protein 8|Complex I 23kD|CI-23kD|NDUFS8|NADH-ubiquinone oxidoreductase 23 kDa subunit|EC 1.6.5.3|mitochondrial|NADH ubiquinone oxidoreductase 23 kDa subunit|EC 1.6.99.3|mitochondrial precursor|Human mitochondrial NADH dehydrogenase ubiquinone Fe S protein 8|CI 23kD|Ubiquinoneiron sulfur protein 8|NDUS8_HUMAN|TYKY
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00217
- O00217
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human NDUFS8
- Fusion protein of Human NDUFS8
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together