Order Entry
Canada
ContactUsLinkComponent
Anti-MT1A Rabbit Polyclonal Antibody (Biotin)
Anti-MT1A Rabbit Polyclonal Antibody (Biotin)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-MT1A Rabbit Polyclonal Antibody (Biotin)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77711-753
  • 77711-754
  • Antigen symbol:
  • MT1A
  • MT1A
  • Antigen synonyms:
  • Metallothionein-IA|Metallothionein2|MT 2|Metallothionein 1S|Metallothionein IA|MT2|MT 1A|MT1A_HUMAN|MT2A|MTC|MT-IA|CES1|Metallothionein II|MT1A|MT1S|MT-1A|MGC32848|MT II|Metallothionein 1A|MT1|Metallothionein-1A|MT IA|MT 2A|Metallothionein 2A|Metallothionein 2|CES 1
  • Metallothionein-IA|Metallothionein2|MT 2|Metallothionein 1S|Metallothionein IA|MT2|MT 1A|MT1A_HUMAN|MT2A|MTC|MT-IA|CES1|Metallothionein II|MT1A|MT1S|MT-1A|MGC32848|MT II|Metallothionein 1A|MT1|Metallothionein-1A|MT IA|MT 2A|Metallothionein 2A|Metallothionein 2|CES 1
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P04731
  • P04731
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Metallothionein-1A protein (1-59AA)
  • Recombinant Human Metallothionein-1A protein (1-59AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together