Anti-LCP2 (phospho-Y128) Rabbit Polyclonal Antibody
Catalog # 77703-028
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:LCP2 (phospho-Y128)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q13094
- Antigen synonyms:76 kDa tyrosine phosphoprotein|SH2 domain containing leukocyte protein of 76kD|SLP 76|SLP-76 tyrosine phosphoprotein|SH2 domain-containing leukocyte protein of 76 kDa|SLP 76 tyrosine phosphoprotein|LCP2|Lymphocyte cytosolic protein 2|SLP76|CG8697|LCP 2|SH2 domain containing leukocyte protein of 76kDa|SH2 domain containing leukocyte protein 76 KD|SLP76 tyrosine phosphoprotein|LCP2_HUMAN|SLP-76
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human SLP-76 around the phosphorylation site of Y128.
- Purification:Antigen affinity purified
- Cat. no.:77703-028
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:5000
Type:
Antigen: LCP2 (phospho-Y128)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat