Anti-KANK2 Rabbit Polyclonal Antibody
Catalog # 77702-510
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:KANK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q63ZY3
- Antigen synonyms:Matrix-remodeling-associated protein 3|SIP|ANKRD25|SRC1-interacting protein|KIAA1518|KN motif and ankyrin repeat domain-containing protein 2|MXRA3|Ankyrin repeat domain-containing protein 25|KANK2|SRC-1-interacting protein|SRC-interacting protein
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human KANK2.
- Purification:Antigen affinity purified
- Cat. no.:77702-510
- Packaging:Vial
Specifications
About this item
IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:40000
Type:
Antigen: KANK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human