Anti-IL4R Rabbit Polyclonal Antibody
Catalog # 77702-927
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:IL4R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:P24394
- Antigen synonyms:IL-4 receptor subunit alpha|IL4-BP|IL 4R alpha|IL-4-binding protein|Soluble IL-4 receptor subunit alpha|IL-4RA|IL-4R-alpha|Il4r|Interleukin 4 receptor|IL-4R subunit alpha|Interleukin 4 receptor alpha chain|IL4RA_HUMAN|IL4R nirs variant 1|IL4RA|Soluble IL-4R-alpha|sIL4Ralpha/prot|Soluble interleukin-4 receptor subunit alpha|CD124
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human IL-4Rα.
- Purification:Antigen affinity purified
- Cat. no.:77702-927
- Packaging:Vial
Specifications
About this item
IHC:1:100-1:300, ELISA:1:40000
Type:
Antigen: IL4R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse