Anti-IGFBP3 (phospho-S183) Rabbit Polyclonal Antibody
77702-906
New Product
- Antigen symbol:IGFBP3 (phospho-S183)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P17936
- Antigen synonyms:IGFBP-3|Binding protein 29|Insulin-like growth factor binding protein 3 precursor|IBP 3|IBP3|IGFBP 3|IGF-binding protein 3|IBP-3|IGF binding protein 3|BP53|Insulin-like growth factor-binding protein 3|BP 53|Acid stable subunit of the 140 K IGF complex|Binding protein 53|Growth hormone dependent binding protein|IBP3_HUMAN|Insulin Like Growth Factor Binding Protein 3|IGFBP3
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human IGFBP3 around the phosphorylation site of S183.
- Purification:Antigen affinity purified
- Cat. no.:77702-906
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:5000
Type:
Antigen: IGFBP3 (phospho-S183)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse