Anti-HARS2 Rabbit Polyclonal Antibody
Catalog # 77687-787
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:HARS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P49590
- Antigen synonyms:HO3|HisRS|HARS related|SYHM_HUMAN|4631412B19Rik|HARSR|Histidine translase|Histidine tRNA ligase|mitochondrial|Histidyl tRNA synthetase 2 mitochondrial|Histidyl tRNA synthetase 2|Histidine tRNA ligase homolog|Histidine--tRNA ligase-like|Probable histidyl-tRNA synthetase|Hars2|HARSL|Histidine tRNA ligase like|Histidine--tRNA ligase|AI593507|Probable histidyl tRNA synthetase mitochondrial|Histidyl tRNA synthetase like
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Probable histidine--tRNA ligase, mitochondrial protein (34-506AA)
- Purification:>95%, Protein G purified
- Cat. no.:77687-787
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200
Type:
Antigen: HARS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse