Anti-GLUD2 Rabbit Polyclonal Antibody
Catalog # 77687-735
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:GLUD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P49448
- Antigen synonyms:GDH2|Glutamate dehydrogenase 2 mitochondrial|DHE4_HUMAN|GDH|Glutamate dehydrogenase 2|mitochondrial|GDH 2|GLUDP1|Glutamate dehydrogenase pseudogene 1|GLUD2
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Glutamate dehydrogenase 2, mitochondrial protein (319-558AA)
- Purification:Antigen affinity purified
- Cat. no.:77687-735
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200
Type:
Antigen: GLUD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat