Anti-Genome polyprotein Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Genome polyprotein
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dengue virus type 1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77712-420
- 77712-419
- Antigen symbol:
- Genome polyprotein
- Genome polyprotein
- Antigen synonyms:
- Non-structural protein 4A|Peptide pr|Envelope protein E|Flavivirin protease NS2B regulatory subunit|EC 2.7.7.48|NS1)|Non-structural protein 2A|Non-structural protein 2B)|RNA-directed RNA polymerase NS5|NS4B)|Non-structural protein 4B|Peptide 2k|EC 2.1.1.57|Flavivirin protease NS3 catalytic subunit|Non-structural protein 3)|EC 3.6.1.15|Protein prM|Capsid protein|EC 3.6.4.13|Non-structural protein 1|Non-structural protein 5)]|Serine protease subunit NS2B|Matrix protein)|Genome polyprotein [Cleaved into: Capsid protein C|Peptide precursor)|Precursor membrane protein)|Core protein)|EC 3.4.21.91|NS4A)|Serine protease NS3|EC 2.1.1.56|NS2A)|Small envelope protein M
- Non-structural protein 4A|Peptide pr|Envelope protein E|Flavivirin protease NS2B regulatory subunit|EC 2.7.7.48|NS1)|Non-structural protein 2A|Non-structural protein 2B)|RNA-directed RNA polymerase NS5|NS4B)|Non-structural protein 4B|Peptide 2k|EC 2.1.1.57|Flavivirin protease NS3 catalytic subunit|Non-structural protein 3)|EC 3.6.1.15|Protein prM|Capsid protein|EC 3.6.4.13|Non-structural protein 1|Non-structural protein 5)]|Serine protease subunit NS2B|Matrix protein)|Genome polyprotein [Cleaved into: Capsid protein C|Peptide precursor)|Precursor membrane protein)|Core protein)|EC 3.4.21.91|NS4A)|Serine protease NS3|EC 2.1.1.56|NS2A)|Small envelope protein M
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Dengue virus type 1
- Dengue virus type 1
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P17763
- P17763
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Dengue virus type 1 Genome polyprotein protein
- Recombinant Dengue virus type 1 Genome polyprotein protein
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together