Anti-CHST6 Rabbit Polyclonal Antibody
77703-130
New Product
- Antigen symbol:CHST6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q9GZX3
- Antigen synonyms:GST4-beta|GlcNAc6ST 5|Carbohydrate sulfotransferase 6|Galactose N acetylglucosamine N acetylglucosamine 6 O sulfotransferase 4 beta|Corneal GlcNAc6-sulfotransferase|Carbohydate sulfotransferase 6|Corneal N acetylglucosamine 6 sulfotransferase|GST4 beta|Gn6st-5|GlcNAc6ST-5|Corneal N-acetylglucosamine-6-O-sulfotransferase|N-acetylglucosamine 6-O-sulfotransferase 5|C GlcNAc6ST|Carbohydrate (N acetylglucosamine 6 O) sulfotransferase 6|CHST6|hCGn6ST|CHST6_HUMAN|C-GlcNAc6ST|Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4-beta
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human CHST6.
- Purification:Antigen affinity purified
- Cat. no.:77703-130
- Packaging:Vial
IF:1:200-1:1000, ELISA:1:20000
Type:
Antigen: CHST6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human