Anti-ATP5L2 Rabbit Polyclonal Antibody
Catalog # 77702-567
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen symbol:ATP5L2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q7Z4Y8
- Antigen synonyms:ATP5MGL|mitochondrial|ATP5K2|ATP5L2Putative ATP synthase subunit g 2|ATPase subunit g 2|ATP synthase membrane subunit g-like protein
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human ATP5L2.
- Purification:Antigen affinity purified
- Cat. no.:77702-567
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IF:1:200-1:1000, ELISA:1:40000
Type:
Antigen: ATP5L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human