Anti-ADAR Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, IHC:1:25-1:100
Type:
Antigen: ADAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77710-828
- 77710-829
- Antigen symbol:
- ADAR
- ADAR
- Antigen synonyms:
- DSRAD_HUMAN|Adenosine deaminase that act on RNA|K88DSRBP|Interferon induced protein 4|Double-stranded RNA-specific editase Adar|IFI4|Double stranded RNA specific adenosine deaminase|Interferon-inducible protein 4|Adar1|dsRNA adenosine deaminase|DRADA|ADAR|AGS6|RNA adenosine deaminase 1|Ifi4 protein|Double-stranded RNA-specific adenosine deaminase|Adenosine deaminase acting on RNA 1 A|136kDa double stranded RNA binding protein|Adenosine deaminase RNA specific 1|AV242451|Dsh|136 kDa double-stranded RNA-binding protein|EC 3.5.4.-|Interferon inducible protein 4|Pre-mRNA adenosine deaminase|Adenosine deaminase RNA specific|IFI-4|RNA-editing deaminase 1|Dsrad|IFI 4|mZaADAR|Adar 1|RNA-editing enzyme 1|G1P1|P136
- DSRAD_HUMAN|Adenosine deaminase that act on RNA|K88DSRBP|Interferon induced protein 4|Double-stranded RNA-specific editase Adar|IFI4|Double stranded RNA specific adenosine deaminase|Interferon-inducible protein 4|Adar1|dsRNA adenosine deaminase|DRADA|ADAR|AGS6|RNA adenosine deaminase 1|Ifi4 protein|Double-stranded RNA-specific adenosine deaminase|Adenosine deaminase acting on RNA 1 A|136kDa double stranded RNA binding protein|Adenosine deaminase RNA specific 1|AV242451|Dsh|136 kDa double-stranded RNA-binding protein|EC 3.5.4.-|Interferon inducible protein 4|Pre-mRNA adenosine deaminase|Adenosine deaminase RNA specific|IFI-4|RNA-editing deaminase 1|Dsrad|IFI 4|mZaADAR|Adar 1|RNA-editing enzyme 1|G1P1|P136
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P55265
- P55265
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human ADAR
- Synthetic peptide of Human ADAR
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together