Order Entry
Canada
ContactUsLinkComponent
Anti-NEK8 Rabbit Polyclonal Antibody
Anti-NEK8 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-NEK8 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77716-579
  • 77716-578
  • Antigen symbol:
  • NEK8
  • NEK8
  • Antigen synonyms:
  • NEK8|NIMA (never in mitosis gene a) related kinase 8|NIMA-related kinase 12a|NEK12A|NIMA-family kinase NEK8|NEK 8|JCK|Nima-related protein kinase 12a|Serine/threonine-protein kinase Nek8|NEK8_HUMAN|Nephrocystin 9|NIMA related kinase 8|Never in mitosis A-related kinase 8|NimA-related protein kinase 8|NPHP9
  • NEK8|NIMA (never in mitosis gene a) related kinase 8|NIMA-related kinase 12a|NEK12A|NIMA-family kinase NEK8|NEK 8|JCK|Nima-related protein kinase 12a|Serine/threonine-protein kinase Nek8|NEK8_HUMAN|Nephrocystin 9|NIMA related kinase 8|Never in mitosis A-related kinase 8|NimA-related protein kinase 8|NPHP9
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q86SG6
  • Q86SG6
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Serine/threonine-protein kinase Nek8 protein (1-300AA)
  • Recombinant Human Serine/threonine-protein kinase Nek8 protein (1-300AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage buffer:
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together