Anti-PRDX3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:5000, WB:1:200-1:1000, IHC:1:25-1:100
Type:
Antigen: PRDX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77716-415
- 77716-414
- Antigen symbol:
- PRDX3
- PRDX3
- Antigen synonyms:
- Antioxidant protein 1|Protein MER5 homolog|PRDX3_HUMAN|AOP 1|AOP1|peroxiredoxin 3|MGC24293|Peroxiredoxin III|AOP-1|MGC104387|SP 22|Thioredoxin dependent peroxide reductase mitochondrial|mitochondrial|PRDX3|SP-22|Thioredoxin-dependent peroxide reductase|PRO1748|SP22|MER5|PRX3|PRX III|HBC189|Prx-III|Peroxiredoxin-3
- Antioxidant protein 1|Protein MER5 homolog|PRDX3_HUMAN|AOP 1|AOP1|peroxiredoxin 3|MGC24293|Peroxiredoxin III|AOP-1|MGC104387|SP 22|Thioredoxin dependent peroxide reductase mitochondrial|mitochondrial|PRDX3|SP-22|Thioredoxin-dependent peroxide reductase|PRO1748|SP22|MER5|PRX3|PRX III|HBC189|Prx-III|Peroxiredoxin-3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P30048
- P30048
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human PRDX3
- Fusion protein of Human PRDX3
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together