Anti-MGEA5 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, WB:1:200-1:1000, IHC:1:25-1:100
Type:
Antigen: MGEA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77716-401
- 77716-402
- Antigen symbol:
- MGEA5
- MGEA5
- Antigen synonyms:
- KIAA0679|N-acetyl-beta-glucosaminidase|OGA_HUMAN|Meningioma-expressed antigen 5|MEA5|Nuclear cytoplasmic O GlcNAcase and acetyltransferase|Mgea5|Hyaluronidase in meningioma|Meningioma expressed antigen 5|HEXC|MGEA 5|N-acetyl-beta-D-glucosaminidase|Beta-N-acetylglucosaminidase|Nuclear cytoplasmic O-GlcNAcase and acetyltransferase|NCOAT|OGA|Protein O-GlcNAcase|Bifunctional protein NCOAT|Beta-hexosaminidase|Beta-N-acetylhexosaminidase
- KIAA0679|N-acetyl-beta-glucosaminidase|OGA_HUMAN|Meningioma-expressed antigen 5|MEA5|Nuclear cytoplasmic O GlcNAcase and acetyltransferase|Mgea5|Hyaluronidase in meningioma|Meningioma expressed antigen 5|HEXC|MGEA 5|N-acetyl-beta-D-glucosaminidase|Beta-N-acetylglucosaminidase|Nuclear cytoplasmic O-GlcNAcase and acetyltransferase|NCOAT|OGA|Protein O-GlcNAcase|Bifunctional protein NCOAT|Beta-hexosaminidase|Beta-N-acetylhexosaminidase
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60502
- O60502
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human MGEA5
- Fusion protein of Human MGEA5
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together