Anti-PNPT1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, WB:1:200-1:1000, IHC:1:50-1:200
Type:
Antigen: PNPT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77716-367
- 77716-366
- Antigen symbol:
- PNPT1
- PNPT1
- Antigen synonyms:
- Polynucleotide phosphorylase 1|PNPASE|3'-5' RNA exonuclease OLD35|PNPT1|Polyribonucleotide nucleotidyltransferase 1|mitochondrial|EC 2.7.7.8|PNPase 1|Polynucleotide phosphorylase-like protein|PNPase old-35
- Polynucleotide phosphorylase 1|PNPASE|3'-5' RNA exonuclease OLD35|PNPT1|Polyribonucleotide nucleotidyltransferase 1|mitochondrial|EC 2.7.7.8|PNPase 1|Polynucleotide phosphorylase-like protein|PNPase old-35
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TCS8
- Q8TCS8
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human PNPT1
- Synthetic peptide of Human PNPT1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together