Anti-GRIA2 Rabbit Polyclonal Antibody
New Product
ELISA:1:2000-1:10000, IHC:1:100-1:300
Type:
Antigen: GRIA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77716-118
- 77716-119
- Antigen symbol:
- GRIA2
- GRIA2
- Antigen synonyms:
- GLURB|AMPA2|GluR-2|GluA2|AMPA-selective glutamate receptor 2|Glutamate receptor ionotropic|GluR-K2|HBGR2|Glutamate receptor 2|GLUR B|AMPA selective glutamate receptor 2|GluR K2|Gria2|GLUR 2|Glutamate receptor ionotropic AMPA 2|GLUR2|AMPA 2|GluR-B|GRIA2_HUMAN
- GLURB|AMPA2|GluR-2|GluA2|AMPA-selective glutamate receptor 2|Glutamate receptor ionotropic|GluR-K2|HBGR2|Glutamate receptor 2|GLUR B|AMPA selective glutamate receptor 2|GluR K2|Gria2|GLUR 2|Glutamate receptor ionotropic AMPA 2|GLUR2|AMPA 2|GluR-B|GRIA2_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P42262
- P42262
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human GRIA2
- Synthetic peptide of Human GRIA2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together