Anti-SHBG Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:5000, IHC:1:25-1:100
Type:
Antigen: SHBG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77716-076
- 77716-077
- Antigen symbol:
- SHBG
- SHBG
- Antigen synonyms:
- Sex hormone-binding globulin|Sex steroid binding protein|Testosterone binding beta globulin|SHBG|Testis-specific androgen-binding protein|Androgen binding protein|Sex steroid-binding protein|SHBG_HUMAN|SBP|Testosterone estradiol binding globulin|TeBG|Testosterone estrogen binding globulin|Testosterone-estrogen-binding globulin|Testosterone-estradiol-binding globulin|Testis specific androgen binding protein|ABP
- Sex hormone-binding globulin|Sex steroid binding protein|Testosterone binding beta globulin|SHBG|Testis-specific androgen-binding protein|Androgen binding protein|Sex steroid-binding protein|SHBG_HUMAN|SBP|Testosterone estradiol binding globulin|TeBG|Testosterone estrogen binding globulin|Testosterone-estrogen-binding globulin|Testosterone-estradiol-binding globulin|Testis specific androgen binding protein|ABP
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P04278
- P04278
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human SHBG
- Synthetic peptide of Human SHBG
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together