Order Entry
Canada
ContactUsLinkComponent
Anti-CCL15 Rabbit Polyclonal Antibody
Anti-CCL15 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-CCL15 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77715-989
  • 77705-612
  • Antigen symbol:
  • CCL15
  • CCL15
  • Antigen synonyms:
  • MIP-1 delta|MRP 2B|HCC2|MIP-5|Mrp2b|NCC 3|HMRP 2B|LKN-1|Small inducible cytokine subfamily A (Cys Cys)|C-C motif chemokine ligand 15|Small inducible cytokine A15|SCYA15|New CC chemokine 3|CCL15(29-92)|CCL15_HUMAN|member 15|MIP1 delta|MIP1d|Chemokine CC-2|MIP 1D|MIP5|Chemokine CC2|LKN1|Macrophage inflammatory protein 5|Mrp-2b|OTTHUMP00000163955|Leukotactin 1|HCC-2|MIP-1D|C-C motif chemokine 15|SY15|NCC3|CC chemokine 3|HMRP2B|Small-inducible cytokine A15|Chemokine (C C motif) ligand 15|NCC-3|Leukotactin-1|SCYL3|CCL15
  • MIP-1 delta|MRP 2B|HCC2|MIP-5|Mrp2b|NCC 3|HMRP 2B|LKN-1|Small inducible cytokine subfamily A (Cys Cys)|C-C motif chemokine ligand 15|Small inducible cytokine A15|SCYA15|New CC chemokine 3|CCL15(29-92)|CCL15_HUMAN|member 15|MIP1 delta|MIP1d|Chemokine CC-2|MIP 1D|MIP5|Chemokine CC2|LKN1|Macrophage inflammatory protein 5|Mrp-2b|OTTHUMP00000163955|Leukotactin 1|HCC-2|MIP-1D|C-C motif chemokine 15|SY15|NCC3|CC chemokine 3|HMRP2B|Small-inducible cytokine A15|Chemokine (C C motif) ligand 15|NCC-3|Leukotactin-1|SCYL3|CCL15
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q16663
  • Q16663
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Synthetic peptide from Human protein at AA range: 51-100
  • Synthetic peptide from Human protein at AA range: 51-100
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage buffer:
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.4.
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.4.
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together