Anti-ZWINT Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:5000, IHC:1:50-1:200
Type:
Antigen: ZWINT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77715-854
- 77715-853
- Antigen symbol:
- ZWINT
- ZWINT
- Antigen synonyms:
- ZWINT1|ZWINT|MGC117174|Human ZW10 interacting protein 1|ZWINT 1|KNTC2AP|KNTC 2 AP|ZW10-interacting protein 1|MGC 117174|HZwint1|HZwint 1|ZW10 interacting protein 1|kinetochore protein|ZW10 interactor|ZW10 interacting kinetochore protein|Zwint-1|ZWINT_HUMAN
- ZWINT1|ZWINT|MGC117174|Human ZW10 interacting protein 1|ZWINT 1|KNTC2AP|KNTC 2 AP|ZW10-interacting protein 1|MGC 117174|HZwint1|HZwint 1|ZW10 interacting protein 1|kinetochore protein|ZW10 interactor|ZW10 interacting kinetochore protein|Zwint-1|ZWINT_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95229
- O95229
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human ZWINT
- Fusion protein of Human ZWINT
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together