Anti-OLFM4 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, WB:1:200-1:1000
Type:
Antigen: OLFM4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77715-559
- 77715-558
- Antigen symbol:
- OLFM4
- OLFM4
- Antigen synonyms:
- OLFM4_HUMAN|GC1|hGC 1|OlfD|bA209J19.1|OLM4|GW112|OLFM4|KIAA4294|Olfactomedin-4|G-CSF-stimulated clone 1 protein|UNQ362|olfactomedin 4|hOLfD|Antiapoptotic protein GW112|hGC-1
- OLFM4_HUMAN|GC1|hGC 1|OlfD|bA209J19.1|OLM4|GW112|OLFM4|KIAA4294|Olfactomedin-4|G-CSF-stimulated clone 1 protein|UNQ362|olfactomedin 4|hOLfD|Antiapoptotic protein GW112|hGC-1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6UX06
- Q6UX06
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human OLFM4
- Fusion protein of Human OLFM4
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together