Anti-KISS1R Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, IHC:1:50-1:200
Type:
Antigen: KISS1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77715-447
- 77715-448
- Antigen symbol:
- KISS1R
- KISS1R
- Antigen synonyms:
- AXOR12|KISS 1 receptor|Metastin receptor|Hypogonadotropin1|KISS 1R|OT7T175|KiSS-1R|Hypogonadotropin 1|KISSR_HUMAN|KiSS-1 receptor|Hypogonadotropin-1|Kiss1r|GPCR54|AXOR 12|GPCR 54|GPR 54|GPR54|Kisspeptins receptor|G-protein coupled receptor 54|hOT7T175|G protein coupled receptor 54|KISS1 receptor|G-protein coupled receptor OT7T175
- AXOR12|KISS 1 receptor|Metastin receptor|Hypogonadotropin1|KISS 1R|OT7T175|KiSS-1R|Hypogonadotropin 1|KISSR_HUMAN|KiSS-1 receptor|Hypogonadotropin-1|Kiss1r|GPCR54|AXOR 12|GPCR 54|GPR 54|GPR54|Kisspeptins receptor|G-protein coupled receptor 54|hOT7T175|G protein coupled receptor 54|KISS1 receptor|G-protein coupled receptor OT7T175
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q969F8
- Q969F8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human KISS1R
- Synthetic peptide of Human KISS1R
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together