Anti-NAPSA Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:5000, IHC:1:50-1:200
Type:
Antigen: NAPSA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77715-206
- 77715-205
- Antigen symbol:
- NAPSA
- NAPSA
- Antigen synonyms:
- Napsin 1|Kidney derived aspartic protease like protein|Napsin A precursor|Napsin-A|NAPA|Kdap|Aspartyl protease 4|KAP|Pronapsin A|NAP1|Napsa|NAPSA_HUMAN|TA01/TA02|SNAPA|napsin A aspartic peptidase|Napsin-1|Asp 4|ASP4
- Napsin 1|Kidney derived aspartic protease like protein|Napsin A precursor|Napsin-A|NAPA|Kdap|Aspartyl protease 4|KAP|Pronapsin A|NAP1|Napsa|NAPSA_HUMAN|TA01/TA02|SNAPA|napsin A aspartic peptidase|Napsin-1|Asp 4|ASP4
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O96009
- O96009
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human NAPSA
- Fusion protein of Human NAPSA
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together