Anti-SLC6A8 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, IHC:1:25-1:100
Type:
Antigen: SLC6A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77714-921
- 77714-920
- Antigen symbol:
- SLC6A8
- SLC6A8
- Antigen synonyms:
- Solute carrier family 6 member 8|CRT|Creatine transporter 1|Sodium and chloride-dependent creatine transporter 1|SLC6A8|Creatine transporter SLC6A8|MGC87396|Solute carrier family 6 (neurotransmitter transporter, creatine), member 8.|Slc6a8|Sodium- and chloride-dependent creatine transporter 1|CT1|CT 1|SC6A8_HUMAN|SLC6A 8|CRTR
- Solute carrier family 6 member 8|CRT|Creatine transporter 1|Sodium and chloride-dependent creatine transporter 1|SLC6A8|Creatine transporter SLC6A8|MGC87396|Solute carrier family 6 (neurotransmitter transporter, creatine), member 8.|Slc6a8|Sodium- and chloride-dependent creatine transporter 1|CT1|CT 1|SC6A8_HUMAN|SLC6A 8|CRTR
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P48029
- P48029
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human SLC6A8
- Synthetic peptide of Human SLC6A8
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together