Anti-PRKDC (phospho-T2647) Rabbit Polyclonal Antibody
77703-861
New Product
- Antigen symbol:PRKDC (phospho-T2647)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:P78527
- Antigen synonyms:DNAPK catalytic subunit|PRKDC|Hyperradiosensitivity complementing 1, mouse, homolog of 1|DNA-PKcs|PRKDC_HUMAN|HYRC 1|Protein Kinase DNA Activated Catalytic Polypeptide|DNA-PK catalytic subunit|XRCC 7|DNA PK catalytic subunit|p460|IMD26|DNPK1|DNA-dependent protein kinase catalytic subunit|Hyper radiosensitivity of murine scid mutation, complementing 1|PKRDC|XRCC7|DNAPK|HYRC1|DNPK 1|DNA dependent protein kinase catalytic subunit|HYRC|p350
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human DNA-PKCS around the phosphorylation site of T2647.
- Purification:Antigen affinity purified
- Cat. no.:77703-861
- Packaging:Vial
IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:10000
Type:
Antigen: PRKDC (phospho-T2647)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human