Anti-GPR87 Rabbit Polyclonal Antibody
77703-695
New Product
- Antigen symbol:GPR87
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9BY21
- Antigen synonyms:GPCR87|GPR95|Gpr87|G-protein coupled receptor 95|G-protein coupled receptor 87|Orphan GPCR 87|GPR 95|KPG 002|Probable G protein coupled receptor 87|MGC131898|hGPCR87|FKSG78|KPG_002|G protein coupled receptor 87|GPR 87|FKSG 78|G protein coupled receptor 95|GPR87_HUMAN
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human GPR87.
- Purification:Antigen affinity purified
- Cat. no.:77703-695
- Packaging:Vial
WB:1:500-1:2000, IF:1:200-1:1000, ELISA:1:5000
Type:
Antigen: GPR87
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse