Order Entry
Canada
ContactUsLinkComponent
Anti-PKD2 (phospho-S812) Rabbit Polyclonal Antibody
Anti-PKD2 (phospho-S812) Rabbit Polyclonal Antibody
Catalog # 77703-665
Supplier:  AFG Bioscience
New Product
undefined
Anti-PKD2 (phospho-S812) Rabbit Polyclonal Antibody
Catalog # 77703-665
Supplier:  AFG Bioscience
Supplier Number:  A39395-100UG
New Product

Specifications

  • Antigen symbol:
    PKD2 (phospho-S812)
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µg
  • Form:
    Liquid
  • Gene ID:
    Q13563
  • Antigen synonyms:
    Polycystic kidney disease 2 protein|Polycystin 2 transient receptor potential cation channel|MGC138468|Autosomal dominant polycystic kidney disease type II protein|Polycystin 2|Polycystwin|MGC138466|Polycystic kidney disease 2 (autosomal dominant)|Autosomal dominant polycystic kidney disease type II|PC2|PKD4|PKD2_HUMAN|TRPP2|PKD 2|Polycystic kidney disease 2|R48321|Polycystin-2|PC 2|APKD2|Transient receptor potential cation channel subfamily P member 2|Polycystin2|PKD2
  • Storage buffer:
    PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
  • Storage temperature:
    Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Synthesized peptide derived from Human PKD2 around the phosphorylation site of S812.
  • Purification:
    Antigen affinity purified
  • Cat. no.:
    77703-665
  • Packaging:
    Vial

Specifications

About this item

IF:1:200-1:1000, ELISA:1:5000

Type:
Antigen: PKD2 (phospho-S812)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat