Anti-CYB5R1 Rabbit Polyclonal Antibody
Catalog # 77703-631
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CYB5R1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9UHQ9
- Antigen synonyms:b5R.1|NADH-cytochrome b5 reductase 1|NAD(P)H:quinone oxidoreductase type 3, polypeptide A2|B5R1|NAD|NB5R1_HUMAN|NQO3A2|Cytochrome b5 reductase 1|EC 1.6.2.2|NADH cytochrome b5 reductase 1|UNQ3049/PRO9865|CYB5R1|B5R2|NAD(P)H:quinone oxidoreductase type 3 polypeptide A2|Humb5R2
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human CYB5R1.
- Purification:Antigen affinity purified
- Cat. no.:77703-631
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: CYB5R1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat