Anti-ZNF699 Rabbit Polyclonal Antibody
Catalog # 77688-932
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ZNF699
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q32M78
- Antigen synonyms:Drosophila|MGC129881|ZNF699|Zinc finger protein 699|hang|MGC129880|ZN699_HUMAN|Hangover homolog|FLJ38144|homolog of|hangover
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Zinc finger protein 699 protein (19-195AA)
- Purification:>95%, Protein G purified
- Cat. no.:77688-932
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200
Type:
Antigen: ZNF699
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human