Anti-TOMM70 Rabbit Polyclonal Antibody
Catalog # 77688-813
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:TOMM70
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Gene ID:O94826
- Antigen synonyms:TOM70|Translocase of outer mitochondrial membrane 70 homolog A|TOM70_HUMAN|Mitochondrial import receptor subunit TOM70|Translocase of outer membrane 70 kDa subunit|Translocase of outer membrane TOM70|FLJ9047|Tomm70a|Mitochondrial precursor proteins import receptor
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Mitochondrial import receptor subunit TOM70 protein (60-340AA)
- Purification:Antigen affinity purified
- Cat. no.:77688-813
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TOMM70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human