Anti-SLC25A4 Rabbit Polyclonal Antibody
Catalog # 77688-659
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:SLC25A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:P12235
- Antigen synonyms:heart/skeletal muscle isoform T1|ATP carrier protein|ADT1_HUMAN|Adenine nucleotide translocator 1 (skeletal muscle)|ADP/ATP translocase 1|SLC25A4|PEO3|ATP carrier protein 1|Solute carrier family 25 member 4|Adenine nucleotide translocator 1|ANT 1|ANT|Adenine Nucleotide Translocase 1|ADP ATP carrier protein heart/skeletal muscle isoform T1|AAC1|ADP|T1|PEO2|ANT1|ADP ATP carrier protein 1
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human ADP/ATP translocase 1 protein (40-74AA)
- Purification:>95%, Protein G purified
- Cat. no.:77688-659
- Packaging:Vial
Specifications
About this item
Type:
Antigen: SLC25A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human