Anti-MYC (phospho-S62) HEK293F cells Recombinant Monoclonal Antibody [clone: 2A10]
Catalog # 77691-320
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:MYC (phospho-S62)
- Clonality:Recombinant Monoclonal
- Clone:2A10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:HEK293F cells
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P01106
- Antigen synonyms:Class E basic helix-loop-helix protein 39|bHLHe39|Proto-oncogene c-Myc|BHLHE39|MYC|Myc proto-oncogene protein|Transcription factor p64
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:A synthesized peptide derived from Human Phospho-MYC (S62)
- Purification:Affinity purified
- Cat. no.:77691-320
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:5000, IF:1:20-1:200
Type:
Antigen: MYC (phospho-S62)
Clonality: Recombinant Monoclonal
Clone: 2A10
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: Rabbit IgG
Reactivity: Human