Anti-IFT46 Rabbit Polyclonal Antibody
100 µg, Vial
Catalog # 77690-770
Supplier: AFG Bioscience
Supplier #: A68408-100UG
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: IFT46
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Size:100 µg
- Packaging:Vial
- Western blot:Yes
- Clonality:Polyclonal
- Purification:>95%, Protein G purified
- Form:Liquid
- Isotype:IgG
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Host:Rabbit
- Shipping temperature:Shipped on blue ice
- Cat. No.:77690-770
- Reactivity:Human|Mouse
- Gene ID:Q9NQC8
- Antigen synonyms:C11orf2|C11orf60Intraflagellar transport protein 46 homolog|IFT46
- Immunogen:Recombinant Human Intraflagellar transport protein 46 homolog protein (1-304AA)
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:IFT46
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



