Anti-NAPG Rabbit Polyclonal Antibody
Catalog # 77690-577
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:NAPG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q99747
- Antigen synonyms:N-ethylmaleimide-sensitive factor attachment protein gamma|Gamma-soluble NSF attachment protein|SNAP-gamma|SNAPG|NAPG
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Gamma-soluble NSF attachment protein (1-312AA)
- Purification:Antigen affinity purified
- Cat. no.:77690-577
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: NAPG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human