Anti-GTF3C6 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GTF3C6
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77725-689
- 77725-690
- Antigen symbol:
- GTF3C6
- GTF3C6
- Antigen synonyms:
- General transcription factor 3C polypeptide 6|TFIIIC 35 kDa subunit|TFIIIC35|C6orf51|NPD020|GTF3C6|CDA020|Transcription factor IIIC subunit 6|Transcription factor IIIC 35 kDa subunit
- General transcription factor 3C polypeptide 6|TFIIIC 35 kDa subunit|TFIIIC35|C6orf51|NPD020|GTF3C6|CDA020|Transcription factor IIIC subunit 6|Transcription factor IIIC 35 kDa subunit
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q969F1
- Q969F1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human General transcription factor 3C polypeptide 6 protein (2-213AA)
- Recombinant Human General transcription factor 3C polypeptide 6 protein (2-213AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together