Anti-KIR3DL3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: KIR3DL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77725-173
- 77690-436
- Antigen symbol:
- KIR3DL3
- KIR3DL3
- Antigen synonyms:
- CD158 antigen-like family member Z|Killer cell inhibitory receptor 1|Killer cell immunoglobulin-like receptor 3DL3|CD158z|KI3L3_HUMAN|KIR3DL3
- CD158 antigen-like family member Z|Killer cell inhibitory receptor 1|Killer cell immunoglobulin-like receptor 3DL3|CD158z|KI3L3_HUMAN|KIR3DL3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N743
- Q8N743
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Killer cell immunoglobulin-like receptor 3DL3 protein (26-322AA)
- Recombinant Human Killer cell immunoglobulin-like receptor 3DL3 protein (26-322AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together