Anti-RPRD1A Rabbit Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: RPRD1A
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77724-976
- 77724-977
- Antigen symbol:
- RPRD1A
- RPRD1A
- Antigen synonyms:
- KIAA4077|CDKN2B-related protein|RPRD1A|Regulation of nuclear pre mRNA domain containing 1A|Cyclin dependent kinase 2B inhibitor related protein|p15INK4B-related protein|MGC19513|FLJ10656|mKIAA4077|Hypothetical protein FLJ10656|RPR1A_HUMAN|Putative uncharacterized protein P15RS|Cyclin dependent kinase inhibitor 2B related protein (p15INK4B related protein)|Cyclin dependent kinase inhibitor related protein|Cyclin-dependent kinase inhibitor 2B-related protein|p15INK4B related protein|Regulation of nuclear pre-mRNA domain-containing protein 1A|Cyclin dependent kinase inhibitor 2B related protein|MGC29396|HsT3101|C77387|RGD1307374|MGC36325
- KIAA4077|CDKN2B-related protein|RPRD1A|Regulation of nuclear pre mRNA domain containing 1A|Cyclin dependent kinase 2B inhibitor related protein|p15INK4B-related protein|MGC19513|FLJ10656|mKIAA4077|Hypothetical protein FLJ10656|RPR1A_HUMAN|Putative uncharacterized protein P15RS|Cyclin dependent kinase inhibitor 2B related protein (p15INK4B related protein)|Cyclin dependent kinase inhibitor related protein|Cyclin-dependent kinase inhibitor 2B-related protein|p15INK4B related protein|Regulation of nuclear pre-mRNA domain-containing protein 1A|Cyclin dependent kinase inhibitor 2B related protein|MGC29396|HsT3101|C77387|RGD1307374|MGC36325
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96P16
- Q96P16
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Regulation of nuclear pre-mRNA domain-containing protein 1A protein (2-276AA)
- Recombinant Human Regulation of nuclear pre-mRNA domain-containing protein 1A protein (2-276AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together