Anti-LRIG1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: LRIG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77724-888
- 77724-887
- Antigen symbol:
- LRIG1
- LRIG1
- Antigen synonyms:
- Leucine-rich repeats and immunoglobulin-like domains protein 1|LRIG1_HUMAN|LIG1|LIG-1|leucine rich repeats and immunoglobulin like domains 1|Leucine rich repeat protein LRIG1|LIG 1|leucine-rich repeats- and immunoglobulin-like domains-cotnaining protein 1|Leucine rich repeat protein|Lrig1|Ortholog of mouse integral membrane glycoprotein LIG 1
- Leucine-rich repeats and immunoglobulin-like domains protein 1|LRIG1_HUMAN|LIG1|LIG-1|leucine rich repeats and immunoglobulin like domains 1|Leucine rich repeat protein LRIG1|LIG 1|leucine-rich repeats- and immunoglobulin-like domains-cotnaining protein 1|Leucine rich repeat protein|Lrig1|Ortholog of mouse integral membrane glycoprotein LIG 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96JA1
- Q96JA1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Leucine-rich repeats and immunoglobulin-like domains protein 1 protein (35-220AA)
- Recombinant Human Leucine-rich repeats and immunoglobulin-like domains protein 1 protein (35-220AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together