Anti-AGR3 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: AGR3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77724-426
- 77724-425
- Antigen symbol:
- AGR3
- AGR3
- Antigen synonyms:
- AGR3_HUMAN|hAG 3|anterior gradient protein 3|Anterior gradient protein 3 homolog|Gm888|AG3|Agr3|PDIA18|Anterior gradient homolog 3|Anterior gradient homolog 3 (Xenopus laevis)|HAG3|Anterior gradient 3|BCMP11|Breast cancer membrane protein 11|hAG-3|AG-3|xenopus|homolog of|Protein disulfide isomerase family A member 18|OTTHUMP00000158512
- AGR3_HUMAN|hAG 3|anterior gradient protein 3|Anterior gradient protein 3 homolog|Gm888|AG3|Agr3|PDIA18|Anterior gradient homolog 3|Anterior gradient homolog 3 (Xenopus laevis)|HAG3|Anterior gradient 3|BCMP11|Breast cancer membrane protein 11|hAG-3|AG-3|xenopus|homolog of|Protein disulfide isomerase family A member 18|OTTHUMP00000158512
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TD06
- Q8TD06
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Anterior gradient protein 3 protein (22-166AA)
- Recombinant Human Anterior gradient protein 3 protein (22-166AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together