Anti-VN1R2 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: VN1R2
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77724-313
- 77724-314
- Antigen symbol:
- VN1R2
- VN1R2
- Antigen synonyms:
- VN1R2_HUMAN|V1r like receptor 2|V1RL2|V1R-like 2|pheromone receptor|vomeronasal 1 receptor 2|VN1R2|V1r-like receptor 2|Vomeronasal type-1 receptor 2|Vomeronasal type 1 receptor 2|G-protein coupled receptor GPCR25|hGPCR25
- VN1R2_HUMAN|V1r like receptor 2|V1RL2|V1R-like 2|pheromone receptor|vomeronasal 1 receptor 2|VN1R2|V1r-like receptor 2|Vomeronasal type-1 receptor 2|Vomeronasal type 1 receptor 2|G-protein coupled receptor GPCR25|hGPCR25
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NFZ6
- Q8NFZ6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Vomeronasal type-1 receptor 2 protein (115-170AA)
- Recombinant Human Vomeronasal type-1 receptor 2 protein (115-170AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together