Anti-Histone H2B (Acetyl-Lys5) Rabbit Polyclonal Antibody
77698-460
New Product
- Form:Liquid
- Gene ID:Q96A08, P33778, P62807
- Antigen synonyms:Histone H2B.f|H2B/f|HIST1H2BB|HIST1H2BA|HIST1H2BC|H2BFK|HIST1H2BG|HIST1H2BI|Testis-specific histone H2B|HIST1H2BE|Histone H2B type 1-A|H2BFH|TSH2B|Histone H2B type 1-B|HIST1H2BF|H2BFA|H2BFG|H2BFL|H2BFF|Histone H2B, testis|Histone H2B.1
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthetic Peptide
- Purification:Affinity purified
- Cat. no.:77698-460
- Packaging:Vial
WB:1:1000-2000
Type:
Antigen: Histone H2B (acetyl-Lys5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat