Anti-Histone H4 (Di-methyl-Lys59) Rabbit Polyclonal Antibody
77698-424
New Product
- Form:Liquid
- Gene ID:P62805
- Antigen synonyms:H4FJ|H4/j|HIST1H4H|H4/I|H4/g|H4FI|H4FA|H4/b|FO108|Histone H4|HIST1H4B|dJ160A22.1|Histone 1 H4h|Histone 1 H4b|HIST1H4E|H4F2|HIST1H4L|H4FC|Histone 1 H4e|H4M|H4/c|H4/e|H4|H4FE|HIST1H4A|H4F2iii|dJ160A22.2|HIST1H4J|dJ221C16.9|Histone 2 H4a|Histone 1 H4f|H4.k|Histone 1 H4a|H4FH|H4FM|H4FB|histone 4 H4|dJ221C16.1|H4/a|HIST1H4K|Histone 1 H4l|H4_HUMAN|H4/p|Histone 1 H4d|H4/k|HIST2H4|H4/d|H4/h|HIST1H4F|HIST1H4D|Histone 1 H4j|H4FG|Histone 1 H4k|Hist4h4|HIST1H4I|H4F2iv|MGC24116|Histone 1 H4c|H4FK|H4/n|HIST1H4C|H4FN|H4FD|Histone 1 H4i|H4/m|HIST2H4A
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthetic Peptide
- Purification:Affinity purified
- Cat. no.:77698-424
- Packaging:Vial
WB:1:500-1:1000, ELISA:1:20000
Type:
Antigen: Histone H4 (Di-methyl-Lys59)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat