Anti-COL1A1 Mouse Monoclonal Antibody
Catalog # 77698-199
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:COL1A1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:P02452
- Antigen synonyms:Collagen alpha-1(I) chain (Alpha-1 type I collagen)
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthetic Peptide of Collagen I
- Purification:Antigen affinity purified
- Cat. no.:77698-199
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC:1:50-1:200
Type:
Antigen: COL1A1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat