Anti-mOrange Mouse Monoclonal Antibody
Catalog # 77698-084
Supplier: AFG Bioscience
New Product
Specifications
- Form:liquid
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Protein
- Purification:Antigen affinity purified
- Cat. no.:77698-084
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:5000
Type:
Antigen: mOrange
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Multiple