Anti-COX4I1 Mouse Monoclonal Antibody
Catalog # 77697-957
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:COX4I1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:liquid
- Gene ID:P13073
- Antigen synonyms:Cytochrome c oxidase subunit 4 isoform 1, mitochondrial|COX IV-1|Cytochrome c oxidase polypeptide IV|COX4|COX4I1|Cytochrome c oxidase subunit IV isoform 1
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Protein
- Purification:Antigen affinity purified
- Cat. no.:77697-957
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:5000
Type:
Antigen: COX4I1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat