Anti-RIOX2 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RIOX2
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77717-284
- 77717-285
- Antigen symbol:
- RIOX2
- RIOX2
- Antigen synonyms:
- Nucleolar protein 52|MINA_HUMAN|MINA|Ribosomal oxygenase MINA|Histone lysine demethylase MINA|ROX|MDIG|MYC induced nuclear antigen|Bifunctional lysine specific demethylase and histidyl hydroxylase MINA|FLJ14393|MYC-induced nuclear antigen|Mineral dust-induced gene protein|60S ribosomal protein L27a histidine hydroxylase|NO52|Myc induced nuclear antigen 53 kDa
- Nucleolar protein 52|MINA_HUMAN|MINA|Ribosomal oxygenase MINA|Histone lysine demethylase MINA|ROX|MDIG|MYC induced nuclear antigen|Bifunctional lysine specific demethylase and histidyl hydroxylase MINA|FLJ14393|MYC-induced nuclear antigen|Mineral dust-induced gene protein|60S ribosomal protein L27a histidine hydroxylase|NO52|Myc induced nuclear antigen 53 kDa
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IUF8
- Q8IUF8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ribosomal oxygenase 2 protein (1-300AA)
- Recombinant Human Ribosomal oxygenase 2 protein (1-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together