Anti-DNAJC10 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DNAJC10
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77717-142
- 77717-143
- Antigen symbol:
- DNAJC10
- DNAJC10
- Antigen synonyms:
- ER-resident protein ERdj5|MTHr|EC 1.8.4.-|Endoplasmic reticulum DNA J domain-containing protein 5|ERdj5|DNAJC10|ERDJ5|UNQ495/PRO1012DnaJ homolog subfamily C member 10|Macrothioredoxin
- ER-resident protein ERdj5|MTHr|EC 1.8.4.-|Endoplasmic reticulum DNA J domain-containing protein 5|ERdj5|DNAJC10|ERDJ5|UNQ495/PRO1012DnaJ homolog subfamily C member 10|Macrothioredoxin
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IXB1
- Q8IXB1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human DnaJ homolog subfamily C member 10 protein (521-793AA)
- Recombinant Human DnaJ homolog subfamily C member 10 protein (521-793AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together