Anti-HIPK1 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HIPK1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77716-917
- 77716-916
- Antigen symbol:
- HIPK1
- HIPK1
- Antigen synonyms:
- Nbak 2|Nbak2|Nuclear body associated kinase 2b|Hipk1|Homeodomain interacting protein kinase 1 like protein|Nuclear body associated kinase 2|HIPK1_HUMAN|Homeodomain-interacting protein kinase 1|Hipk 1|Homeodomain interacting protein kinase 1|Myak
- Nbak 2|Nbak2|Nuclear body associated kinase 2b|Hipk1|Homeodomain interacting protein kinase 1 like protein|Nuclear body associated kinase 2|HIPK1_HUMAN|Homeodomain-interacting protein kinase 1|Hipk 1|Homeodomain interacting protein kinase 1|Myak
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86Z02
- Q86Z02
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Homeodomain-interacting protein kinase 1 protein (1007-1151AA)
- Recombinant Human Homeodomain-interacting protein kinase 1 protein (1007-1151AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together