Anti-NFATC2IP Rabbit Polyclonal Antibody
Catalog # 77704-818
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:NFATC2IP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q8NCF5
- Antigen synonyms:NFATC2-interacting protein|NIP45|Nuclear factor of activated T-cells, cytoplasmic 2-interacting protein|nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2 interacting protein|45 kDa NFAT-interacting protein|RAD60|NFAT-interacting protein, 45-KD|Nfatc2ip|ESC2|NF2IP_HUMAN|Nuclear factor of activated T-cells|45 kDa NF-AT-interacting protein|cytoplasmic 2-interacting protein
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human NFATc2IP
- Purification:Antigen affinity purified
- Cat. no.:77704-818
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:10000
Type:
Antigen: NFATC2IP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human